Learn More
Invitrogen™ Acétate de sodium (3 M), pH 5,5, sans RNase
Description
Invitrogen Molecular biology grade, 3M Sodium Acetate solution is supplied in one bottle containing 100 mL. The solution is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Invitrogen's nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.
Order Info
Conditions d’expédition : Température ambiante
Spécifications
Spécifications
| Nom ou substance chimique | Acétate de sodium |
| Contenu et stockage | Entreposez le produit à température ambiante. |
| Concentration | 3 M |
| À utiliser avec (application) | Applications de l’ARN |
| Type d’emballage | Flacon |
| pH | 5.5 |
| Gamme de produit | Ambion |
| Pureté | Grade de biologie moléculaire |
| Quantité | 100 mL |
| Shipping condition (Condition d'expédition) | Température ambiante |
| Afficher plus de résultats |
Usage exclusivement réservé à la recherche. Ne pas utiliser dans des procédures de diagnostic.