missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Single Place Magnetic Stand (for one 1.5 mL tube)

Catalog No. AM10026
Encompass
Change view
Click to view available options
Quantity:
1 Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM10026 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM10026 Supplier Invitrogen™ Supplier No. AM10026
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

Accommodates one 1.5 mL microfuge tube.

• Single Tube Magnetic Stand accommodates one 1.5 mL microfuge tube
• With this stand, centrifugation is replaced by the attraction between capture beads in a solution and the magnet in the stand, resulting in the beads collecting on one side of the microfuge tube.
• Thus, the captured species (e.g., mRNA) is quickly and efficiently separated from the solution's other components.

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage Store at room temperature.
For Use With (Equipment) Magnetic Stand
Shipping Condition Room Temperature
Product Line Ambion
Quantity 1 Each
Type Magnetic Stand
Will Ambion magnetic stands (AM10050 & AM10026) work for 0.2 mL individual PCR tubes or strips?

Ambion magnetic stands (AM10050 & AM10026) are not designed for and do not work well for 0.2 mL individual PCR tubes or strips.

For Research Use Only. Not for use in diagnostic procedures.