missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNaseAlert™ QC System v2

Catalog No. 4479769
Encompass
Change view
Click to view available options
Quantity:
480 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4479769 480 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4479769 Supplier Invitrogen™ Supplier No. 4479769

missing translation for 'validMainframeMsg'

The RNaseAlert™ QC System v2 detects RNase activity in convenient and sensitive fluorimetric assay that delivers results in real time.

The RNaseAlert™ QC System v2 detects RNase activity in convenient and sensitive fluorimetric assay that delivers results in real time. This version contains substrate that is ∼60% more sensitive than previous version. The kit contains sufficient reagents for 5 x 96 (480) high-throughput assays.

  • Detects as little as approx. 0.3 pg of RNase A with fluorometer
  • Simple, straight-forward, 30-minute assay
  • Designed to monitor RNase activity in column fractions during protein purificatio
  • Use with DNaseAlert™ QC System to quantitatively detect both RNases and DNases in same sample
  • Compatible with DNaseAlert QC System

Specifications

Content And Storage

Store at -20°C:
• 5 tubes of RNaseAlert™ Substrate v2
• 5 mL 10X RNaseAlert Lab Test Buffer
• 500 μL RNase A

Store at room temperature:
• 250 mL RNaseZap™ Solution

Store at any temperature:
• 50 mL Nuclease-free Water

Quantity 480 reactions
Reagent Type RNase Control Reagent
Product Line RNaseAlert
For Use With (Application) RNA Extraction

For Research Use Only. Not for use in diagnostic procedures.