missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNaseAlert™ Lab Test Kit v2

Catalog No. 4479768
Encompass
Change view
Click to view available options
Quantity:
25 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4479768 25 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4479768 Supplier Invitrogen™ Supplier No. 4479768

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

RNaseAlert™ Lab Test Kit v2 detects RNase activity in a convenient and sensitive fluorimetric assay that delivers results in real time.

RNaseAlert™ Lab Test Kit v2 detects RNase activity in a convenient and sensitive fluorimetric assay that delivers results in real time. This kit is suitable for testing small numbers of sample and ensuring that solutions, tubes, tips, etc. in your lab are RNase-free. This new version contains a more sensitive substrate that is ∼60% more sensitive than the previous version. The kit contains sufficient reagents for 25 reactions.

  • Detects as little as approx. 0.3 pg of RNase A with a fluorometer
  • Simple, straight-forward 30-minute assay

Specifications

Content And Storage

Store at -20°C:
• 25 tubes of RNaseAlert™ Substrate v2
• 250 μL 10X RNaseAlert Lab Test Buffer
• 500 μL RNase A

Store at room temperature:
• 50 mL RNaseZap™ Solution

Store at any temperature:
• 1.75 mL Nuclease-free Water

Quantity 25 reactions
Reagent Type RNase Control Reagent
Product Line RNaseAlert
For Use With (Application) RNA Extraction

For Research Use Only. Not for use in diagnostic procedures.