missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNaseZap™ RNase Decontamination Wipes Refill

Invitrogen RNaseZap™ Wipes are RNaseZap™ in a convenient towel format. You simply wipe RNases off of surfaces and rinse with RNase-free water.

$163.00 - $404.00

Specifications

Format Wipes
Content And Storage Store at room temperature.
Shipping Condition Room Temperature
Reagent Type RNase Control Reagent
Product Line Ambion, ElectroZap
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Product Type Price Quantity & Availability  
AM9786
Encompass
View Documents Promotion Details
Invitrogen™
AM9786
100 Sheets Pop-up dispenser, 100 wipes
Each for $163.00
Only null left
 
AM9788
Encompass
View Documents Promotion Details
Invitrogen™
AM9788
300 Sheets Refill, 3 x 100 wipes on rolls
Each for $404.00
Only null left
 
Description

Description

Invitrogen RNaseZap™ Wipes are RNaseZap™ in a convenient towel format. You simply wipe RNases off of surfaces and rinse with RNase-free water. Supplied as 100 pop-up sheets in a single container.

• Convenient towel format
• Completely removes RNase contamination from glass and plastic surfaces Ideal for cleaning work surfaces, pipettors, and equipment that must be RNase-free

Works immediately on contact; no baking or autoclaving necessary The popular Invitrogen™ RNaseZap™ is now provided in a convenient wipe format. RNaseZap™ Wipes are designed to eliminate RNase contamination on work surfaces, pipettors and equipment that must be RNase-free. RNaseZap™ is a formulation of three ingredients known to be active against RNase. It effectively removes even high levels of RNase contamination that similar products cannot. With thorough rinsing, RNaseZap™ leaves no residues that are inhibitory to enzymatic reactions.

Note: The components are packaged separately. The wipes and RNaseZap™ Solution are loaded into the pop-up dispenser prior to first use.

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Wipes
Room Temperature
Ambion, ElectroZap
Store at room temperature.
RNase Control Reagent
Videos
SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.