Learn More
Invitrogen™ RNaseZap™ RNase Decontamination Wipes Refill
Description
Invitrogen™ RNaseZap Wipes are RNaseZap™ in a convenient towel format. You simply wipe RNases off of surfaces and rinse with RNase-free water. Supplied as 3 rolls of 100 sheets plus solution to refill existing pop-up dispenser.
- Convenient towel format
- Completely removes RNase contamination from glass and plastic surfaces Ideal for cleaning work surfaces, pipettors, and equipment that must be RNase-free
- Works immediately on contact; no baking or autoclaving necessary
The popular Invitrogen™ RNaseZap™ is now provided in a convenient wipe format. RNaseZap™ Wipes are designed to eliminate RNase contamination on work surfaces, pipettors and equipment that must be RNase-free. RNaseZap™ is a formulation of three ingredients known to be active against RNase. It effectively removes even high levels of RNase contamination that similar products cannot. With thorough rinsing, RNaseZap™ leaves no residues that are inhibitory to enzymatic reactions.
Note: The components are packaged separately. The wipes and RNaseZap™ Solution are loaded into the pop-up dispenser prior to first use.
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Format | Wipes |
| Product Type | Pop-up dispenser, 100 wipes |
| Content And Storage | Store at room temperature. |
| Shipping Condition | Room Temperature |
| Quantity | 100 Sheets |
| Reagent Type | RNase Control Reagent |
| Product Line | Ambion, ElectroZap |
Frequently Asked Questions (FAQs)
The formulation for RNaseZAP RNase Decontamination Solution (Cat. Nos. AM9782, AM9784, AM9780) is proprietary. The pH is 7.0-7.4.
For Research Use Only. Not for use in diagnostic procedures.