missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNaseZap™ RNase Decontamination Wipes Refill

Catalog No. AM9788
Encompass
Change view
Click to view available options
Quantity:
100 Sheets
300 Sheets
Product Type:
Pop-up dispenser, 100 wipes
Refill, 3 x 100 wipes on rolls
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
AM9788 300 Sheets Refill, 3 x 100 wipes on rolls
AM9786 100 Sheets Pop-up dispenser, 100 wipes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AM9788 Supplier Invitrogen™ Supplier No. AM9788

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Invitrogen RNaseZap™ Wipes are RNaseZap™ in a convenient towel format. You simply wipe RNases off of surfaces and rinse with RNase-free water.

Invitrogen™ RNaseZap Wipes are RNaseZap™ in a convenient towel format. You simply wipe RNases off of surfaces and rinse with RNase-free water. Supplied as 3 rolls of 100 sheets plus solution to refill existing pop-up dispenser.

  • Convenient towel format
  • Completely removes RNase contamination from glass and plastic surfaces Ideal for cleaning work surfaces, pipettors, and equipment that must be RNase-free
  • Works immediately on contact; no baking or autoclaving necessary

The popular Invitrogen™ RNaseZap™ is now provided in a convenient wipe format. RNaseZap™ Wipes are designed to eliminate RNase contamination on work surfaces, pipettors and equipment that must be RNase-free. RNaseZap™ is a formulation of three ingredients known to be active against RNase. It effectively removes even high levels of RNase contamination that similar products cannot. With thorough rinsing, RNaseZap™ leaves no residues that are inhibitory to enzymatic reactions.

Note: The components are packaged separately. The wipes and RNaseZap™ Solution are loaded into the pop-up dispenser prior to first use.

Order Info

Shipping Condition: Room temperature

Specifications

Format Wipes
Product Type Refill, 3 x 100 wipes on rolls
Content And Storage Store at room temperature.
Shipping Condition Room Temperature
Quantity 300 Sheets
Reagent Type RNase Control Reagent
Product Line Ambion, ElectroZap
What is the formulation and pH of RNaseZAP Rnase Decontamination Solution (Cat. Nos. AM9782, AM9784, AM9780)?

The formulation for RNaseZAP RNase Decontamination Solution (Cat. Nos. AM9782, AM9784, AM9780) is proprietary. The pH is 7.0-7.4.

For Research Use Only. Not for use in diagnostic procedures.