missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNase AWAY™ Decontamination Reagent

Catalog No. 10328011
Encompass
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10328011 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10328011 Supplier Invitrogen™ Supplier No. 10328011

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ready-to-use solution for eliminating RNase and DNA contamination from labware.

  • Ready-to-use solution for eliminating RNase and DNA contamination from labware
  • Apply evenly over surface of glassware or plasticware to be treated and then rinse it away with distilled water
  • Unwanted RNase and DNA contamination are eliminated
  • Nonabrasive, noncarcinogenic, and nonbiologically corrosive

Order Info

Shipping Condition: Room temperature

Specifications

Format Liquid
Content And Storage Store at room temperature.
Shipping Condition Room Temperature
Quantity 250 mL
Reagent Type RNase Control Reagent
Product Line RNase Away
For Use With (Application) RNA Extraction
What is the recommended contact duration time of the RNase AWAY Decontamination Reagent (Cat. No. 10328011) with the according surface which has to be decontaminated, before rinsing?

There is no recommended time period for the RNase AWAY Decontamination Reagent (Cat. No. 10328011) to be in contact with the surface which has to be decontaminated. You do not need to leave the product act for a given time. Just apply it and rub with a wipe. After that you can rinse it with water.

Can I use RNAse AWAY Decontamination Reagent to get rid of DNA contamination?

RNase AWAY Decontamination Reagent is a ready-to-use solution for eliminating RNase and DNA contamination from labware. Apply it evenly over the surface of glassware or plasticware to be treated and then rinse it away with distilled water.

What quality testing is performed on your RNase AWAY Decontamination Reagent (Cat. No. 10328011)?

RNase AWAY Decontamination Reagent is functionally tested for the elimination of RNase activity and DNA. No detectable RNase activity or DNA is observed.

For Research Use Only. Not for use in diagnostic procedures.