missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Poly(A)Purist™ MAG Kit

Catalog No. AM1922M
Encompass
Change view
Click to view available options
Product Type:
MAG Kit
MAG Kit with Manual
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
AM1922M MAG Kit with Manual
AM1922 MAG Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AM1922M Supplier Invitrogen™ Supplier No. AM1922M

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Store Nuclease–free Water, Microfuge Tubes and Microfuge tubes with Spin Columns at room temperature. Store 2X Binding Solution, Wash Solution 1, Wash Solution 2 and Oligo(dT) Cellulose at 4°C. Store RNA Storage Solution, 5M Ammonium Acetate and Glycogen(5mg/mL) at -20°C.

Requires

Magnetic stand

Ambion™ Poly(A)Purist™ MAG Kit (patent pending) uses magnetic technology for the selection of mRNA that typically requires only a single round of selection yet yields RNA that is more highly enriched and devoid of rRNA than other procedures.

Ambion™ Poly(A)Purist™ MAG Kit (patent pending) uses magnetic technology for the selection of mRNA that typically requires only a single round of selection yet yields RNA that is more highly enriched and devoid of rRNA than other procedures. Each kit contains sufficient reagents for the isolation of mRNA from up to 8 mg of total RNA.

  • Highest recovery of mRNA of any available kit
  • High stringency for lowering residual rRNA content
  • Single round of selection sufficient for most applications

The superior performance of Poly(A)Purist™ MAG Kit is clearly demonstrated by a comparison of the mRNA recovered using 4 other kits (see Figure). The kit uses batch binding of RNA to premeasured aliquots of oligo(dT) cellulose to circumvent the problems of slow flow rates and clogged columns often experienced during conventional, gravity-driven chromatography. The magnetic bead–based selection employed in the delivers increased processing speed, ease of handling, and scalability. The entire selection procedure can be completed in as little as 45 minutes (see Figure).

Accessory Products:
A magnetic stand is required for use this kit. They are available in several formats, including the Single Place Magnetic Stand (Cat. No. AM10026) and the 6 Tube Magnetic Stand (Cat. No. AM10055).

User Guide Alert (July 2023):
Step 3 of the Preparation of Oligo (dT) MagBeads section (page 7) of the User Guide is misleading. Please follow this instruction for Step 3: add an equivalent mass of Oligo (dT) Magbeads into the selected empty tube and then continue with Oligo (dT) Magbeads preparation according to steps 4-9.

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage Store Nuclease-free Water, Microfuge Tubes and Microfuge tubes with Spin Columns at room temperature. Store 2X Binding Solution, Wash Solution 1, Wash Solution 2 and Oligo(dT) Cellulose at 4°C. Store RNA Storage Solution, 5M Ammonium Acetate and Glycogen(5 mg/mL) at -20°C.
Format Kit
Isolation Technology Magnetic Bead, Oligo dT Affinity
Sample Type RNA
Final Product Type mRNA
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) Microarray Analysis, Reverse Transcriptase PCR (RT-PCR), Microinjection, Nucleic Acid Labeling, In Vitro Translation, cDNA Library Construction, Northern Blotting, Nuclease Protection Assays
Product Line Ambion, Poly(A)Purist
Product Type MAG Kit with Manual
Quantity 1 kit
Target mRNA
Test Time 45 min.
Show More Show Less
What is the expected yield of mRNA from total RNA?

mRNA yield is usually 1 to 3% of the total RNA. For example, if you begin with 100 µg of total RNA, you should expect a yield of 1 to 3 µg of mRNA.

For Research Use Only. Not for use in diagnostic procedures.