missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ mMESSAGE mMACHINE™ T7 ULTRA Transcription Kit

Catalog No. AMB13455
Encompass
Change view
Click to view available options
Quantity:
50 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AMB13455 50 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AMB13455 Supplier Invitrogen™ Supplier No. AMB13455

missing translation for 'validMainframeMsg'

Generate RNA transcripts with higher protein yields

The mMESSAGE mMACHINE T7 Ultra Kit combines new cap analog, anti-reverse cap analog (ARCA), with patented high-yield transcription technology to generate RNA transcripts that produce higher protein yields compared to other transcripts upon translation.

  • RNA Yield: 20-30μg
  • Synthesize more protein from in vitro-transcribed RNA
  • Express RNA transcripts more efficiently both in vitro and in vivo
  • Stabilize transcripts in vivo with included reagents for poly(A) tailing

Specifications

Content And Storage T7 Enzyme Mix, 10X Reaction Buffer 2XNTP⁄ARCA Solution, GTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, E-PAP, 5X E-PAP Buffer, ATP Solution, MnCl2, and Formaldehyde Load Dye are all stored at –20°C. Nuclease-free Water may be stored at any temperature.
Final Product Type ARCA Capped Synthetic mRNA, mRNA, Synthetic mRNA
Format Kit
Sample Type mRNA, Synthetic mRNA
For Use With (Application) In Vitro Transcription, synthetic mRNA, mRNA, transfection, LNPs, GENEart, linearized DNA plasmid, delivery, PCR products
No. of Reactions 50 Reactions
Product Line Ambion, mMESSAGE mMACHINE
Product Type T7 In Vitro Transcription kit
Quantity 50 Reactions
Promoter T7
Shipping Condition Dry Ice
Yield 20-30 μg
Starting Material PCR Products, Linearized DNA plasmid
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.