missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E. coli Total RNA

Catalog No. AM7940
Encompass
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM7940 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM7940 Supplier Invitrogen™ Supplier No. AM7940

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The highest-quality RNA available from Invitrogen, it is DNase-treated and subjected to unsurpassed quality control standards. One tube containing 200 μg is provided at a concentration of 1 mg/mL.

The highest-quality RNA available from Invitrogen, it is DNase-treated and subjected to unsurpassed quality control standards. One tube containing 200 μg is provided at a concentration of 1 mg/mL. The samples are processed using Invitrogen RNA isolation reagents to produce highly pure, intact RNA. A stringent DNase treatment is performed to ensure that the RNA is ready for use in any downstream application, including RT-PCR. Because it is impossible to remove every DNA molecule from an RNA sample, each preparation is tested to ensure that any residual DNA contamination is insignificant. The integrity of the RNA is verified by capillary electrophoresis using the Agilent™ 2100 bioanalyzer.

Order Info

Shipping Condition: Dry ice

Specifications

Bacterial Content RNA: varies
Concentration 1 mg/mL
Content And Storage 200 μg E. coli Total RNA; store at or below –70°C

Description E. coli total RNA isolated from DH5α cells
Final Product Type E. coli total RNA stored in THE RNA Storage Solution (1 mM Sodium Citrate, pH 6.4; Cat No. AM7000)
For Use With (Application) RT-PCR, ribonuclease protection assays, S1 nuclease assays, cDNA synthesis, Northern analysis, IVT
Quantity 200 μg
Sample Treatment DNase-Treated
Shipping Condition Dry Ice
Do you offer isolated, purified RNA?

Yes, please visit this website (http://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/rna-extraction/rna-types/purified-rna.html) for our FirstChoice total RNA isolated from human tissues, as well as from E. coli and human cell lines.

For Research Use Only. Not for use in diagnostic procedures.