missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GlycoBlue™ Coprecipitant (15 mg/mL)

Catalog No. AM9516
Encompass
Change view
Click to view available options
Quantity:
300 μL
5 x 300 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM9516 5 x 300 μL
AM9515 300 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AM9516 Supplier Invitrogen™ Supplier No. AM9516

GlycoBlue Coprecipitant consists of a blue dye covalently linked to glycogen, a branched chain carbohydrate, which is useful as a nucleic acid coprecipitant.

GlycoBlue Coprecipitant consists of a blue dye covalently linked to glycogen, a branched chain carbohydrate, which is useful as a nucleic acid coprecipitant. The attached dye increases visibility of the pellet. This product is an ideal coprecipitant in nuclease protection assays at 1/100 dilution of stock solution.

Features of GlycoBlue™ Coprecipitant:
• Ideal for RT-PCR
• Increases pellet mass and visibility
• Quantitative recovery of low concentrations (ng/mL) of nucleic acid
• Prevents pellet loss in nuclease protection assays

Order Info

Shipping Condition: Dry Ice

Specifications

Concentration 15 mg/mL
Content And Storage Store at –20°C.
Packaging Type Tube
Product Line Ambion, GlycoBlue
Purity Molecular Biology Grade
Quantity 5 x 300 μL
Shipping Condition Dry Ice
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.