Learn More
Invitrogen™ Formamide (Deionized)
Ambion formamide has been deionized, is nuclease-free, and comes packaged as a liquid in an amber glass bottle under argon.
$215.00 - $864.00
Specifications
| Color | Colorless |
|---|---|
| Content And Storage | Store at 4° or -20°C as small aliquots. |
| Packaging Type | Bottle |
| Product Type | Formamide |
| Product Line | Ambion |
Description
Ambion formamide has been deionized, is nuclease-free, and comes packaged as a liquid in an amber glass bottle under argon. Supplied in 5 bottles containing 500 g each. Formamide lowers the melting temperature of nucleic acid duplexes and is widely used in applications such as Southern and northern blotting, nuclease protection assays, S1 mapping, and in situ hybridization. For critical applications, it is recommended that the formamide from previously opened bottles be deionized before use.
Appearance: Clear, colorless
Molecular Weight: 45.04
Purity: ≥99.5%
Quality Control: The product has been tested for RNase and DNase contamination.
Order Info
Shipping Condition: Room temperature
Specifications
| Colorless | |
| Bottle | |
| Ambion | |
| Liquid |
| Store at 4° or -20°C as small aliquots. | |
| Formamide | |
| >99.5% |
For Research Use Only. Not for use in diagnostic procedures.