missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ElectroZap™ Electrode Decontamination Solution

Catalog No. AM9785
Encompass
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM9785 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM9785 Supplier Invitrogen™ Supplier No. AM9785

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Invitrogen™ ElectroZap is a non-corrosive, stable cleaning solution that has been specially formulated to clean electrode surfaces in asimple, quick, and efficient manner and is effective against high levels of RNase or DNase contamination.

Invitrogen™ ElectroZap is a non-corrosive, stable cleaning solution that has been specially formulated to clean electrode surfaces in asimple, quick, and efficient manner and is effective against high levels of RNase or DNase contamination. Researchers performing RNA analysis often require the use of pH meter to prepare their RNase-free solutions. The use of same pH meter by several laboratory personnel, however, can often lead to problems with RNase contamination, especially when preparing solutions that cannot be DEPC-treated or autoclaved. Special precautions must be taken when removing RNase contamination from pH meter so that the accuracy will be unaffected and there is no residue left by cleaning solution. Simply spray ElectroZap directly onto the electrode and then rinse off with RNase-free water — no residue that may interfere with subsequent enzymatic reactions remains. Supplied in 250 mL bottle.

  • Easily rinsed away leaving no residue
  • Stable, safe, and non-corrosive
  • No need for strong acid or alkali
  • Also removes DNases and other proteins

Order Info

Shipping Condition: Room temperature

Specifications

Format Liquid
Content And Storage Store ElectroZap™ at room temperature.
Shipping Condition Room Temperature
Quantity 250 mL
Reagent Type RNase Control Reagent
Product Line Ambion, ElectroZap
For Use With (Application) RNA Extraction

For Research Use Only. Not for use in diagnostic procedures.