missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cells-to-Signal™ Lysis Buffer (10 ml Bottle)

Catalog No. AM8728
Encompass
Change view
Click to view available options
Quantity:
5 x 10 mL bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM8728 5 x 10 mL bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM8728 Supplier Invitrogen™ Supplier No. AM8728

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The same lysis buffer found in the Invitrogen Cells-to-Signal™ Kit. Supplied in five 10 mL bottles.

The same lysis buffer found in the Invitrogen Cells-to-Signal™ Kit. Supplied in five 10 mL bottles.

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Lysis Buffer may be stored at any temperature.
Product Line Ambion, Cells-to-Signal
Product Type Cell Lysis Buffer
Quantity 5 x 10 mL bottle
Shipping Condition Room Temperature
When should I use Cells-to-Signal lysis vs. purified RNA for the Clariom GO Screen Assay?

Generally, the more samples you are running for the Clariom GO Screen Assay, the more time and cost will be saved with direct Cells-to-Signal lysis.

Which cell lines can I lyse with the Cells-to-Signal Kit for use with the Clariom GO Screen Assay?

Two cell lines have been fully verified for the Clariom GO Screen assay, HepG2 and HeLa. General guidance for creating lysates with other cell lines is included in the user guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0018542_ClariomGO_Screen384HT_UG.pdf).

How do I order the Clariom GO Screen Assay?

You can purchase the Clariom GO Screen Assay (Cat. No. 952361). To start a GO Screen project, contact your local representative and/or complete the quote request form via the product page (https://www.thermofisher.com/order/catalog/product/952361). Our verified service providers are available to help you run your GO Screen assay and can assist with the management of your project.

For Research Use Only. Not for use in diagnostic procedures.