missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Amberlite™ XAD4, 20-60 mesh

Catalog No. 111015791
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015791 1 kg
111015790 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015791 Supplier MilliporeSigma™ Supelco™ Supplier No. XAD41KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Hydrophobic polyaromatic resin

Nonionic macroreticular resin that adsorbs and releases ionic species through hydrophobic and polar interactions; usually used under isocratic conditions.

Specifications

Particle Size 20/60 Mesh
Pore Size 100 Å
Packaging Poly Bottle
Quantity 1 kg
Surface Area (Metric) 750 m2/g
Product Line Amberlite
MDL Number MFCD00132704
Density 1.08 g/mL (skeletal)(lit.)
CAS 37380-42-0

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow