missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ XAD-1180, polymeric adsorbent

Catalog No. AC299155000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC299155000 500 g Plastic Bottle
AC299151000 100 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC299155000 Supplier Thermo Scientific Chemicals Supplier No. 299155000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 9003-69-4 | C10H10

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9003-69-4
Molecular Formula C10H10
Molecular Weight (g/mol) 0.00
MDL Number MFCD00132704
InChI Key MYRTYDVEIRVNKP-UHFFFAOYSA-N
Synonym divinylbenzene, 1,2-divinylbenzene, o-divinylbenzene, benzene, diethenyl, benzene, 1,2-diethenyl, divinylbenzen, divinyl benzen, divinyl-benzene, poly divinylbenzene, divinylbenzene dvb
PubChem CID 66666
SMILES *

Specifications

Physical Form Beads
Particle Size Particle size analysis: uniformity coefficient: <1.80, harmonic mean size: 0.350 to 0.600mm, <0.250mm: <5.0%,>0.850mm: <10.0%
Packaging Plastic Bottle
Water Retention Capacity moisture holding capacity: 61.0 to 67.0%
Color White to Yellow
Merck Index 14, 382
Quantity 500 g
IUPAC Name 1,2-bis(ethenyl)benzene
Chemical Name or Material Amberlite™ XAD 1180

RUO – Research Use Only