missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ IRC-120, H form, ion-exchange resin

Catalog No. AC466712500
Change view
Click to view available options
Quantity:
250 g
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466712500 250 g
AC466710050 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466712500 Supplier Thermo Scientific Chemicals Supplier No. 466712500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 65098-55-7

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 65098-55-7
MDL Number MFCD00132707

Specifications

Quantity 250 g
Solubility Information Solubility aq. soln.: insoluble.
Chemical Name or Material Amberlite* IRC-120
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Causes serious eye damage.

Precautionary Statement GHS P Statement Wear eye protection/face protection.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Immediately call a POISON CENTER or doctor/physician.

RUO – Research Use Only