missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Amberlite™ IRA-900 Chloride Form

Catalog No. 111015869
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015869 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015869 Supplier MilliporeSigma™ Supelco™ Supplier No. 216585500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Amberlite™ IRA-900 chloride form is a macroporous, polystyrenic type 1 resin.

Amberlite™ IRA-900 chloride form resin was found to catalyze the photodecomposition of carbon tetrachloride in ethanol.

Specifications

Packaging Poly Bottle
pH 0 to 14
Quantity 500 g
Product Line Amberlite
Capacity (Metric) 1.0 meq/mL by wetted bed volume
MDL Number MFCD00132712
CAS 9050-97-9

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow