missing translation for 'onlineSavingsMsg'
Learn More

Amberlite IRA-410(Cl), ion exchange resin

Catalog No. AAA1773436
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1773436 500 g
AAA1773422 100 g
AAA177340E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1773436 Supplier Thermo Scientific Chemicals Supplier No. A1773436

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 9002-26-0

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9002-26-0
MDL Number MFCD00132710

Specifications

Odor Amine-like
Merck Index 14,382
Quantity 500 g
Chemical Name or Material Amberlite IRA-410(Cl), ion exchange resin
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only