Learn More
Fisherbrand™ Amber Wide Mouth Packer Glass Bottles with White Polypropylene Caps
Amber, USP Type III soda-lime glass ideal for storage of light sensitive samples or dry powders. Easy removal of contents.
$88.00 - $1,025.64
Specifications
| Color | Amber |
|---|---|
| Material | Glass (Type 3) |
| Closure Color | White |
| Closure Included | Yes |
| Mouth | Wide |
| Catalog Number | Capacity (Metric) | Closure Liner | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Capacity (Metric) | Closure Liner | Price | Quantity & Availability | |||||
|
FB02911799A
|
15 mL | Pulp/Vinyl |
Case of 624 Each for $1,025.64
|
|
|||||
|
FB02911438
|
30 mL | PTFE/Polyethylene Foam |
Case of 24 Each for $91.36
|
|
|||||
|
FB02911854
|
2500 mL | PTFE/Polyethylene Foam |
Case of 12 Each for $363.40
|
|
|||||
|
FB02911440
|
120 mL | PTFE/Polyethylene Foam |
Case of 24 Each for $88.00
|
|
|||||
|
FB02911443
|
950 mL | PTFE/Polyethylene Foam |
Case of 12 Each for $146.64
|
|
|||||
|
FB02911796
|
1250 mL | PTFE/Polyethylene Foam |
Case of 6 Each for $107.72
|
|
|||||
|
FB02911439
|
60 mL | PTFE/Polyethylene Foam |
Case of 24 Each for $93.16
|
|
|||||
|
FB02911441
|
250 mL | PTFE/Polyethylene Foam |
Case of 24 Each for $137.64
|
|
|||||
|
FB02911442
|
500 mL | PTFE/Polyethylene Foam |
Case of 12 Each for $107.12
|
|
|||||
|
FB02911448A
|
60 mL | Pulp/Vinyl |
Case of 216 Each for $428.36
|
|
|||||
|
FB02911857A
|
8 mL | Pulp/Vinyl |
Case of 912 Each for $977.08
|
|
|||||
Description
- Wide opening for easy transfer of contents
- Amber color filters out ultraviolet wavelengths, ideal for light sensitive solids
- Classic shape of vitamin bottles, often used in pharmaceutical packaging
- Often used in chemical and environmental applications
- Caps offer good impact strength and pliability, can be remelted after they are formed
- Cap liner is made of a vinyl coating applied to high density polyethylene coated paper and then it is laminated to pulpboard
Specifications
| Amber | |
| White | |
| Wide | |
| Polypropylene |
| Glass (Type 3) | |
| Yes | |
| Wide Mouth Packer Bottle |