Learn More
Electron Microscopy Sciences AMB Passive Air Balance Platform, 45 x 30 cm, Stainless Steel Surface
Numéro de catalogue. 501899198
Encompass
These high performance anti vibration platform for balances are weighted to 25kg and are for use with light instruments such as balances. They support loads to 120kg and are available in three sizes, 45x30cm, 40x50cm and 60x60cm. Their small footprint also makes them ideal for use when space is at a minimum. Working height is 6cm.
Size: 45 x 30 cm
Surface: Stainless Steel
Type: Pneumatic air
Numéro de catalogue. 501899198 Fournisseur Electron Microscopy Sciences Code fournisseur 6640SS
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
These high performance anti vibration platform for balances are weighted to 25kg and are for use with light instruments such as balances. They support loads to 120kg and are available in three sizes, 45x30cm, 40x50cm and 60x60cm. Their small footprint also makes them ideal for use when space is at a minimum. Working height is 6cm.
Size: 45 x 30 cm
Surface: Stainless Steel
Type: Pneumatic air