missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AM 80

Catalog No. 350710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
350710 10 mg
350750 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 350710 Supplier Tocris Bioscience™ Supplier No. 3507/10

RARα agonist; anticancer agent

Retinoic acid receptor α (RARα) agonist that induces differentiation (ED50 = 0.79 nM) and apoptosis of HL-60 cells in vitro. Exhibits antiproliferative effects against a variety of human tumor cells lines (mean values of 35, 40 and 60% growth inhibition at 0.1, 1 and 10 μM respectively) and displays anticancer activity against acute promyelocytic leukemia in vivo.

Chemical Identifiers

CAS 94497-51-5
Molecular Formula C22H25NO3

Specifications

Quantity 10 mg
Formula Weight 351.44
Purity >99%
Molecular Weight (g/mol) 351.45
Chemical Name or Material AM 80
Recommended Storage Store at Room Temperature