CAS RN 94497-51-5
IUPAC Name:
4-[(5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)carbamoyl]benzoic acid
Synonyms:
tamibarotene amnoid tamibaro retinobenzoic acid
Molecular Weight (g/mol):
351.45
Molecular Formula:
C22H25NO3
InChi Key:
MUTNCGKQJGXKEM-UHFFFAOYSA-N
SMILES:
CC1(C)CCC(C)(C)C2=CC(NC(=O)C3=CC=C(C=C3)C(O)=O)=CC=C12
1
–
2
of
2
results
1
Tamibarotene
CAS: 94497-51-5 Molecular Formula: C22H25NO3 Molecular Weight (g/mol): 351.45 MDL Number: MFCD00866188 InChI Key: MUTNCGKQJGXKEM-UHFFFAOYSA-N Synonym: tamibarotene,retinobenzoic acid,4-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl carbamoyl benzoic acid,amnolake,tamibaro,amnoid,unii-08v52gz3h9,4-5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl carbamoyl benzoic acid,chembl25202 PubChem CID: 108143 ChEBI: CHEBI:32181 IUPAC Name: 4-[(5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl)carbamoyl]benzoic acid SMILES: CC1(C)CCC(C)(C)C2=CC(NC(=O)C3=CC=C(C=C3)C(O)=O)=CC=C12
2
R&D Systems™ AM 80
CAS: 94497-51-5 Molecular Formula: C22H25NO3