missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Aluminum Sign: HEARING PROTECTION REQUIRED IN THIS AREA. HEARING DAMAGE CAN OCCUR.

Catalog No. 19111632
Change view
Click to view available options
Dimensions (L x W):
14 x 10 in.
16 x 16 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19111632 14 x 10 in.
19111630 16 x 16 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19111632 Supplier Brady™ Supplier No. 144131
Only null left

Improve workplace safety with proper signage and markings. This Brady™ aluminum sign is a durable, clear and effective tool to raise awareness to workplace hazards.

  • Aluminum material is abrasion-resistant and chemical-resistant
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • Aluminum signs provide the strength and durability of metal, but never rust and can tolerate extreme temperatures, years of chemical spills, wash downs and cleanings
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use

Specifications

Material Aluminum
Shape Rectangular
Dimensions (L x W) 14 x 10 in.
Displayed Text HEARING PROTECTION REQUIRED IN THIS AREA. HEARING DAMAGE CAN OCCUR.
Color Blue