Learn More
Brady™ Aluminum Sign: STAY ALERT AND YIELD TO FORKLIFT TRAFFIC
Description
- Aluminum material is abrasion-resistant and chemical-resistant
- Safety signs are important facility visuals that are vital to communicating warnings and other safety information
- Aluminum signs provide the strength and durability of metal, but never rust and can tolerate extreme temperatures, years of chemical spills, wash downs and cleanings
- Bold and bright, providing high visibility to critical messages
- For indoor and outdoor use
Specifications
Specifications
| Material | Aluminum |
| Shape | Rectangular |
| Dimensions (L x W) | 23.875 x 6 in. |
| Displayed Text | STAY ALERT AND YIELD TO FORKLIFT TRAFFIC |
| Color | Yellow on White |