missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, Aerosol Refractory Paint, Thermo Scientific Chemicals

Catalog No. AA40391CLC
Change view
Click to view available options
Quantity:
13 oz.
5 x 13 oz.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40391CLC 13 oz.
AA403910CL 5 x 13 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA40391CLC Supplier Thermo Scientific Chemicals Supplier No. 040391CLC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Provides easy application of uniform, thin, protective high temperature Aluminum oxide coatings. Substrates can be ceramic, graphite or metal. Aluminum oxide is also widely used as an abrasive. In the paper-making process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Provides easy application of uniform, thin, protective high temperature Al2O3 coatings
  • Substrates can be ceramic, graphite or metal

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
IUPAC Name dialuminium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Boiling Point -44°C
Color White
Quantity 13 oz.
Flash Point -104°C (-155°F)
Packaging Aerosol Spray Can
UN Number UN1950
Solubility Information Slightly soluble in water. insoluble in diethyl ether Practically insoluble in ethanol
Chemical Name or Material Aluminum oxide, Aerosol Refractory Paint
Hazard Category H222-H319-H336-H350-X
Hazard Statement GHS H Statement
H222-H319-H336
Extremely flammable aerosol.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P304+P340-P305+P351+P338-P308+P313-P501c
DOTInformation Transport Hazard Class: 2.1; Packaging Group: ; Proper Shipping Name: AEROSOLS
TSCA Yes
Recommended Storage Ambient temperatures

For Manufacturing and Laboratory Use. Not for beverage use