missing translation for 'onlineSavingsMsg'
Learn More

Aluminum ICP Standard, 1,000 ppm Al in 3% HCl, Ricca Chemical

Supplier:  Ricca Chemical Company PAL1KH500

Catalog No. PAL1KH500


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Aluminum ICP Standard
7784-27-2 , 7647-01-0 , 7429-90-5 , 7732-18-5
97.0,4.59,0.1
AlH18N3O18
< 1
100°C
Miscible with water
Standard
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]
375.129
24567
1mL = 1mg Al (1, 000ppm Al)Al in 3% HCl
Natural Poly Bottle
93.0,4.41,0.09
Liquid
Colorless
0.0°C
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Calibration Standard
SWCIQHXIXUMHKA-UHFFFAOYSA-N
aluminum;trinitrate;nonahydrate
500 mL
Laboratory

Chemical Identifiers

7784-27-2
375.129
24567
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]
AlH18N3O18
SWCIQHXIXUMHKA-UHFFFAOYSA-N
aluminum;trinitrate;nonahydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 18 months

SDS
Documents

Documents

Promotions

Promotions