missing translation for 'onlineSavingsMsg'
Learn More

Aluminum fluoride trihydrate, 97%

Catalog No. AA1149622
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1149622 100 g
AA1149636 500 g
AA11496A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1149622 Supplier Thermo Scientific Chemicals Supplier No. 01149622

CAS: 15098-87-0 | AlF3H6O3 | 138.022 g/mol

Aluminum fluoride trihydrate is used as an additive for the production of aluminum by electrolysis. It is also used to inhibit fermentation and in the preparation of low index films. It serves as a flux ingredient for the removal of magnesium in the refining of aluminum scrap. Further, it is used in the manufacture of aluminum silicates, ceramics and glass. In addition, it acts as a catalyst for organic synthesis and filler in glass industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15098-87-0
Molecular Formula AlF3H6O3
Molecular Weight (g/mol) 138.022
MDL Number MFCD00003426
InChI Key BKLZIAYVINRQEJ-UHFFFAOYSA-K
Synonym aluminum trifluoride trihydrate, aluminum 3+ trihydrate trifluoride, aluminum fluoride-water 1/3/3, aluminum fluoride, trihydrate,, aluminum 3+ ion trihydrate trifluoride, fluoride, fluoride, aluminum salt, hydrate, hydrate, hydrate, fluoride
PubChem CID 22002992
IUPAC Name aluminum;trifluoride;trihydrate
SMILES O.O.O.[F-].[F-].[F-].[Al+3]

Specifications

Melting Point 250°C (in -3H2O)
Color White
Physical Form Powder
Quantity 100 g
Merck Index 14,339
Solubility Information Slightly soluble in water,acids and alkalis. Insoluble in alcohol and acetone.
Formula Weight 138.02 (83.98 Anhydrous)
Percent Purity 97%
Odor Odorless
Density 2.17 g/mL
Chemical Name or Material Aluminum fluoride trihydrate
Show More Show Less
Hazard Category H315-H320-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 232-051-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only