missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Chloride, 6 N, Ricca Chemical

$239.90 - $239.90

Chemical Identifiers

CAS 7784-13-6
Molecular Formula AlCl3H12O6
Molecular Weight (g/mol) 241.42
MDL Number MFCD00149134
InChI Key JGDITNMASUZKPW-UHFFFAOYSA-K
Synonym Aluminum (III) Chloride Hexahydrate
PubChem CID 24564
ChEBI CHEBI:30115
IUPAC Name aluminium(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
570-4
SDS View Documents
Ricca Chemical Company
5704
120 mL Natural Poly Bottle
Each for $239.90
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7784-13-6
241.42
JGDITNMASUZKPW-UHFFFAOYSA-K
24564
aluminium(3+) hexahydrate trichloride
AlCl3H12O6
MFCD00149134
Aluminum (III) Chloride Hexahydrate
CHEBI:30115
O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]

Specifications

Colorless to Green
32.17,66.0,0.13
AlCl3H12O6
Aluminum (III) Chloride Hexahydrate
O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]
241.42
CHEBI:30115
Natural Poly Bottle
120 mL
6Normal
7784-13-6 , 7732-18-5 , 7440-44-0
33.48,68.0,0.14
MFCD00149134
JGDITNMASUZKPW-UHFFFAOYSA-K
aluminium(3+) hexahydrate trichloride
24564
Laboratory
2.5 to 3.5
Liquid
Aluminum Chloride
Videos
SDS
Documents

Documents