missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Chloride, 6 N, Ricca Chemical

Catalog No. 5704
Change view
Click to view available options
Quantity:
120 mL
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
5704 120 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5704 Supplier Ricca Chemical Company Supplier No. 5704

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7784-13-6 , 7732-18-5 , 7440-44-0
Molecular Formula AlCl3H12O6
Molecular Weight (g/mol) 241.42
MDL Number MFCD00149134
InChI Key JGDITNMASUZKPW-UHFFFAOYSA-K
Synonym Aluminum (III) Chloride Hexahydrate
PubChem CID 24564
ChEBI CHEBI:30115
IUPAC Name aluminium(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]

Specifications

Color Colorless to Green
CAS Min % 32.17, 66.0, 0.13
CAS Max % 33.48, 68.0, 0.14
Grade Laboratory
Packaging Natural Poly Bottle
pH 2.5 to 3.5
Quantity 120 mL
Physical Form Liquid
Concentration or Composition Notes 6Normal
Chemical Name or Material Aluminum Chloride