missing translation for 'onlineSavingsMsg'
Learn More

Aluminum AA Standard, 1000 ppm Al in 3% HNO3, Ricca Chemical

Catalog No. AAL1KN100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1KN100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1KN100 Supplier Ricca Chemical Company Supplier No. AAL1KN100

Chemical Identifiers

CAS 7784-27-2 , 7697-37-2 , 7784-27-2 , 7732-18-5
Molecular Formula AlH18N3O18
Molecular Weight (g/mol) 375.129
InChI Key SWCIQHXIXUMHKA-UHFFFAOYSA-N
PubChem CID 24567
IUPAC Name aluminum;trinitrate;nonahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Al
CAS Min % 93.0, 4.17, 1.29
CAS Max % 96.0, 4.34, 1.34
Equivalence 1 mL = 1 mg Al
Physical Form Liquid
Color Colorless
pH < 2
Melting Point Approximately 0°C
Boiling Point Approximately 100°C
For Use With (Equipment) Atomic Absorption Spectrophotometer
Solubility Miscible with water
Solution Type Calibration Standard
Type Single-element Standard Solution
Quantity 100 mL
Grade Laboratory
Chemical Name or Material Aluminum AA Standard
Show More Show Less