missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum AA Standard, 1000 ppm Al in 3% HCl, Ricca Chemical
$152.83 - $369.63
Chemical Identifiers
| CAS | 7784-27-2 |
|---|---|
| Molecular Formula | AlH18N3O18 |
| Molecular Weight (g/mol) | 375.129 |
| InChI Key | SWCIQHXIXUMHKA-UHFFFAOYSA-N |
| PubChem CID | 24567 |
| IUPAC Name | aluminum;trinitrate;nonahydrate |
| SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AAL1KH-100
|
Ricca Chemical Company
AAL1KH100 |
100 mL | Natural LDPE Bottle |
Each for $152.83
|
|
||||
|
AAL1KH-500
|
Ricca Chemical Company
AAL1KH500 |
500 mL | Natural Poly Bottle |
Each for $369.63
|
|
||||
Chemical Identifiers
| 7784-27-2 | |
| 375.129 | |
| 24567 | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3] |
| AlH18N3O18 | |
| SWCIQHXIXUMHKA-UHFFFAOYSA-N | |
| aluminum;trinitrate;nonahydrate |
Specifications
| Natural LDPE Bottle | |
| 93.0,4.41,0.09 | |
| 97.0,4.59,0.1 | |
| Liquid | |
| Colorless | |
| SWCIQHXIXUMHKA-UHFFFAOYSA-N | |
| aluminum;trinitrate;nonahydrate | |
| 100 mL | |
| Aluminum AA Standard |