missing translation for 'onlineSavingsMsg'
Learn More

Aluminum AA Standard, 1000 ppm Al in 3% HCl, Ricca Chemical

Catalog No. AAL1KH100
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAL1KH100 100 mL Natural LDPE Bottle
AAL1KH500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1KH100 Supplier Ricca Chemical Company Supplier No. AAL1KH100

Chemical Identifiers

CAS 7784-27-2 , 7647-01-0 , 7429-90-5 , 7732-18-5
Molecular Formula AlH18N3O18
Molecular Weight (g/mol) 375.129
InChI Key SWCIQHXIXUMHKA-UHFFFAOYSA-N
PubChem CID 24567
IUPAC Name aluminum;trinitrate;nonahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Al
CAS Min % 93.0, 4.41, 0.09
CAS Max % 97.0, 4.59, 0.1
Equivalence 1 mL = 1 mg Al
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Quantity 100 mL
Chemical Name or Material Aluminum AA Standard