missing translation for 'onlineSavingsMsg'
Learn More

Aluminon

Catalog No. AC103060250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC103060250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC103060250 Supplier Thermo Scientific Chemicals Supplier No. 103060250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 569-58-4 | C22H23N3O9 | 473.438 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 569-58-4
Molecular Formula C22H23N3O9
Molecular Weight (g/mol) 473.438
MDL Number MFCD00040925
InChI Key AIPNSHNRCQOTRI-UHFFFAOYSA-N
Synonym Aurintricarboxylic acid, triammonium salt
PubChem CID 54729869
ChEBI CHEBI:87398
IUPAC Name triazanium;5-[(3-carboxy-4-oxidophenyl)-(3-carboxy-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]-2-oxidobenzoate
SMILES C1=CC(=C(C=C1C(=C2C=CC(=O)C(=C2)C(=O)O)C3=CC(=C(C=C3)[O-])C(=O)[O-])C(=O)O)[O-].[NH4+].[NH4+].[NH4+]

Specifications

Melting Point 220.0°C to 225.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Suitability suitable for determination of aluminum
Beilstein 10, IV, 4161
Merck Index 15, 318
Color Red
Solubility Information Solubility in water: 800g/L (20°C). Other solubilities: slightly soluble in ethanol, insoluble in petroleum ether
Quantity 25 g
Physical Form Crystalline Powder
Formula Weight 473.44
Chemical Name or Material Aluminon
Show More Show Less
What is the solubility of Aluminon?

Aluminon is soluble in water at a concentration of ~800 g/L at 20°C. It is slightly soluble in ethanol and insoluble in petroleum ether.

Hazard Category Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only