missing translation for 'onlineSavingsMsg'
Learn More

Aluminum standard solution, for AAS, 1 mg/ml Al in 2 to 5% HNO3

Catalog No. AC195805000
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC195805000 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC195805000 Supplier Thermo Scientific Chemicals Supplier No. 195805000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7784-27-2 | AlH18N3O18 | 375.129 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For atomic adsorption spectroscopy

Specifications

Chemical Name or Material Aluminum standard solution
Name Note 1 mg/ml Al in 0.5N HNO3
Packaging Plastic bottle
CAS 7784-27-2, 7697-37-2, 7732-18-5
Density 1.0200g/mL
Specific Gravity 1.02
Physical Form Powder
Molecular Formula AlH18N3O18
Concentration 990 to 1010 μg/mL of Al(3+) in 2 to 5% HNO3
MDL Number MFCD00134029
Color Yellow
Melting Point -3.0°C
Merck Index 15, 347
Boiling Point 101.0°C
Solubility Soluble in water
InChI Key SWCIQHXIXUMHKA-UHFFFAOYSA-N
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]
IUPAC Name aluminum;trinitrate;nonahydrate
Molecular Weight (g/mol) 375.129
Quantity 500 mL
PubChem CID 24567
Formula Weight 375.13
Grade AAS
Show More Show Less

RUO – Research Use Only