missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, for chromatography, basic, Brockmann I, 50-200 μm, 90A

Catalog No. AC463350010
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC463350010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC463350010 Supplier Thermo Scientific Chemicals Supplier No. 463350010

CAS: 1344-28-1 | Al2O3 | 101.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
IUPAC Name dialuminium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Boiling Point 2977.0°C
Melting Point 2030.0°C
Quantity 1 kg
Linear Formula Al2O3
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: slightly sol. in alkaline sol.(forming hydroxides), practically insoluble in non-polar organic solvent, difficultly soluble in mineral acids
Formula Weight 101.96
Grade Chromatography
Specific Gravity 3.97
Density 3.9700 g/mL
Chemical Name or Material Aluminum oxide
EINECSNumber 215-691-6

RUO – Research Use Only