missing translation for 'onlineSavingsMsg'
Learn More

Alternate Metals 1 For ICP, SPEX CertiPrep™

Catalog No. WP11500
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Triple-leached LDPE bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
WP11500 500 mL Triple-leached LDPE bottle
WP11 125 mL Triple-leached LDPE bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. WP11500 Supplier SPEX CertiPrep Group LLC Supplier No. WP11500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Al20μg/mL
Fe20μg/mL
V20μg/mL
Co10μg/mL
Cu10μg/mL
Mn10μg/mL
Ni10μg/mL
Zn10μg/mL
Be5μg/mL
Sb5μg/mL
Tl5μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix 2% HNO3
Industry Type Wastewater Testing
Quantity 500 mL
Grade Environmental
Concentration or Composition (by Analyte or Components) Components in 2% HNO3:
Al    •20μg/mL
Sb    •5μg/mL
Be    •5μg/mL
Co    •10μg/mL
Cu    •10μg/mL
Fe    •20μg/mL
Mn    •10μg/mL
Ni    •10μg/mL
Tl    •5μg/mL
V    •20μg/mL
Zn    •10μg/mL
Chemical Name or Material Alternate Metals 1, For ICP/AA Alternate Water Pollution and Wastewater Standard
Hazard Category Causes severe skin burns and eye damage Causes serious eye damage May cause cancer.
DOTInformation Hazardous
Recommended Storage Ambient

Product intended for laboratory use only