missing translation for 'onlineSavingsMsg'
Learn More

Alphazurine A, MP Biomedicals™

Catalog No. ICN15473025
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15473025 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15473025 Supplier MP Biomedicals Inc Supplier No. 0215473025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Soluble in water (1mg/mL); Store at Room Temperature (15° to 30°C)

Chemical Identifiers

CAS 3486-30-4
Molecular Formula C37H35N2NaO6S2
Molecular Weight (g/mol) 690.805
MDL Number MFCD00044440
InChI Key FTUYQIPAPWPHNC-UHFFFAOYSA-M
Synonym acid blue 7, alphazurine a, patent blue a, fenazo bluexg, erio glaucine x, hidacid blue a, carmine blue af, hidacid blue af, kiton blue a, patent blue af
PubChem CID 62498
IUPAC Name sodium;4-[[4-[benzyl(ethyl)amino]phenyl]-[4-[benzyl(ethyl)azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]benzene-1,3-disulfonate
SMILES CCN(CC1=CC=CC=C1)C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC=CC=C4)C=C3)C5=C(C=C(C=C5)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+]

Specifications

Color Purple
Quantity 25 g
Formula Weight 690.8
Physical Form Powder
Chemical Name or Material Alphazurine A