Learn More
Ansell AlphaTec™ 58-530B Liquid-Proof, Chemical Resistant Glove
Chemical protective and oil-resistant waterproof gauntlet gloves offer a confident grip and a comfortable inner lining. Ideal for outdoor applications.
$58.46 - $58.46
Specifications
ANSI Abrasion Rating ANSI 5
Certifications/Compliance 21 CFR 170-199
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, Category lll, EN 1149
Chlorinated No
Color Gray, Teal
View More Specs
Products 5
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17-999-821
View Documents
Ansell
58530B070
7
Pack of 6 Each for $58.46
Case of 12 Pack for $701.52
Only null left
17-999-822
View Documents
Ansell
58530B080
8
Pack of 6 Each for $58.46
Case of 12 Pack for $669.60
Only null left
17-999-823
View Documents
Ansell
58530B090
9
Pack of 6 Each for $58.46
Case of 12 Pack for $669.60
Only null left
17-999-824
View Documents
Ansell
58530B100
10
Pack of 6 Each for $58.46
Case of 12 Pack for $669.60
Only null left
17-999-825
View Documents
Ansell
58530B110
11
Pack of 6 Each for $58.46
Case of 12 Pack for $669.60
Only null left
Description
Description
  • Combination of liquid proof chemical resistance and grip, with flexibility and dexterity
  • Reliable chemical protection for added user safety.
  • ANSELL GRIP™ technology ensures that the coating polymer does not penetrate the liner during manufacture
  • Coating is consistent and sound, significantly reducing the likelihood of chemical leakage onto the skin
  • Recommended for outdoor applications
Specifications
Specifications
ANSI 5
EN 420:2003 + A1:2009, Category lll, EN 1149
Gray, Teal
Nitrile
No
JKT
Gauntlet
Acrylic
AlphaTec™ 58-530B
29 to 31 cm
21 CFR 170-199
No
Yes
EN 388:3122A, EN 388:4121
Type A
Bacteria, Fungi only
ANSELL GRIP™ Technology
Lined
11 to 12 in.
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'