missing translation for 'onlineSavingsMsg'
Learn More

alpha-(Trichloromethyl)benzyl Acetate 98.0+%, TCI America™

Catalog No. T312225G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T312225G 25 g
T3122250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T312225G Supplier TCI America Supplier No. T312225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 90-17-5
Molecular Formula C10H9Cl3O2
Molecular Weight (g/mol) 267.53
MDL Number MFCD00000804
InChI Key JKRWZLOCPLZZEI-UHFFFAOYSA-N
Synonym Acetic Acid alpha-(Trichloromethyl)benzyl Ester, 2,2,2-Trichloro-1-phenylethyl Acetate, Acetic Acid 2,2,2-Trichloro-1-phenylethyl Ester
PubChem CID 7007
IUPAC Name (2,2,2-trichloro-1-phenylethyl) acetate
SMILES CC(=O)OC(C1=CC=CC=C1)C(Cl)(Cl)Cl

Specifications

Melting Point 88°C
Color White
Boiling Point 282°C
Quantity 25 g
Formula Weight 267.53
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material alpha-(Trichloromethyl)benzyl Acetate
EINECSNumber (3)-1023
RTECSNumber AJ8375000
TSCA Yes