missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Terpineol 95.0+%, TCI America™
Chemical Identifiers
| CAS | 98-55-5 |
|---|---|
| Molecular Formula | C10H18O |
| Molecular Weight (g/mol) | 154.253 |
| MDL Number | MFCD00001557 |
| InChI Key | WUOACPNHFRMFPN-UHFFFAOYSA-N |
| Synonym | alpha-terpineol, terpineol, p-menth-1-en-8-ol, 2-4-methylcyclohex-3-en-1-yl propan-2-ol, dl-alpha-terpineol, 1-p-menthen-8-ol, 1-menthene-8-ol, terpineol schlechthin, carvomenthenol, terpenol |
| PubChem CID | 17100 |
| ChEBI | CHEBI:22469 |
| IUPAC Name | 2-(4-methylcyclohex-3-en-1-yl)propan-2-ol |
| SMILES | CC1=CCC(CC1)C(C)(C)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
T098425ML
|
N/A | 25 mL |
N/A
|
|
|||||
|
T0984100ML
|
N/A | 100 mL |
N/A
|
|
|||||
|
T0984500ML
|
N/A | 500 mL |
N/A
|
|
|||||
Chemical Identifiers
| C10H18O | |
| MFCD00001557 | |
| alpha-terpineol, terpineol, p-menth-1-en-8-ol, 2-4-methylcyclohex-3-en-1-yl propan-2-ol, dl-alpha-terpineol, 1-p-menthen-8-ol, 1-menthene-8-ol, terpineol schlechthin, carvomenthenol, terpenol | |
| CHEBI:22469 | |
| CC1=CCC(CC1)C(C)(C)O |
Specifications
| 98-55-5 | |
| 218°C | |
| MFCD00001557 | |
| WUOACPNHFRMFPN-UHFFFAOYSA-N | |
| 2-(4-methylcyclohex-3-en-1-yl)propan-2-ol | |
| 17100 | |
| 154.25 | |
| Crystalline Lumps |
| White-Yellow | |
| C10H18O | |
| alpha-terpineol, terpineol, p-menth-1-en-8-ol, 2-4-methylcyclohex-3-en-1-yl propan-2-ol, dl-alpha-terpineol, 1-p-menthen-8-ol, 1-menthene-8-ol, terpineol schlechthin, carvomenthenol, terpenol | |
| CC1=CCC(CC1)C(C)(C)O | |
| 154.253 | |
| CHEBI:22469 | |
| ≥95.0% (GC) | |
| alpha-Terpineol |
Safety and Handling
EINECSNumber : (3)-2323
RTECSNumber : WZ6700000
TSCA : Yes