missing translation for 'onlineSavingsMsg'
Learn More

alpha-Terpineol 95.0+%, TCI America™

Supplier:  TCI America T0984100ML

Quantity 100 mL

 View more versions of this product

Catalog No. T0984100ML

Only null left

Description

Description

Specifications

Chemical Identifiers

98-55-5
154.253
WUOACPNHFRMFPN-UHFFFAOYSA-N
17100
2-(4-methylcyclohex-3-en-1-yl)propan-2-ol
C10H18O
MFCD00001557
alpha-terpineol, terpineol, p-menth-1-en-8-ol, 2-4-methylcyclohex-3-en-1-yl propan-2-ol, dl-alpha-terpineol, 1-p-menthen-8-ol, 1-menthene-8-ol, terpineol schlechthin, carvomenthenol, terpenol
CHEBI:22469
CC1=CCC(CC1)C(C)(C)O

Specifications

alpha-Terpineol
White-Yellow
C10H18O
MFCD00001557
WUOACPNHFRMFPN-UHFFFAOYSA-N
2-(4-methylcyclohex-3-en-1-yl)propan-2-ol
17100
154.25
Crystalline Lumps
98-55-5
218°C
100 mL
alpha-terpineol, terpineol, p-menth-1-en-8-ol, 2-4-methylcyclohex-3-en-1-yl propan-2-ol, dl-alpha-terpineol, 1-p-menthen-8-ol, 1-menthene-8-ol, terpineol schlechthin, carvomenthenol, terpenol
CC1=CCC(CC1)C(C)(C)O
154.253
CHEBI:22469
≥95.0% (GC)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2323

RTECSNumber : WZ6700000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions