accessibility menu, dialog, popup

UserName

CH3(R)CH3H3COH

CAS RN 98-55-5

IUPAC Name: 2-[(1R)-4-methylcyclohex-3-en-1-yl]propan-2-ol
Synonyms: (+)-α-terpineol fema number 3045 lily of valley α-terpineol, sodium salt terpenol α-terpinenol α-terpineole α-terpilenol sodium salt α-terpineol (r)-(+)-α-terpineol
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: WUOACPNHFRMFPN-VIFPVBQESA-N
SMILES: CC1=CC[C@@H](CC1)C(C)(C)O
Percent Purity 
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (8)
  • (8)
Physical Form 
  • (3)
  • (10)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (3)
  • (2)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (2)
  • (3)
  • (5)
  • (8)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (8)
  • (8)
  • (3)
  • (10)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)