missing translation for 'onlineSavingsMsg'
Learn More

alpha-Terpineol 95.0+%, TCI America™

Catalog No. T0984500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0984500ML 500 mL
T098425ML 25 mL
T0984100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T0984500ML Supplier TCI America Supplier No. T0984500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 98-55-5
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00001557
InChI Key WUOACPNHFRMFPN-UHFFFAOYSA-N
Synonym alpha-terpineol, terpineol, p-menth-1-en-8-ol, 2-4-methylcyclohex-3-en-1-yl propan-2-ol, dl-alpha-terpineol, 1-p-menthen-8-ol, 1-menthene-8-ol, terpineol schlechthin, carvomenthenol, terpenol
PubChem CID 17100
ChEBI CHEBI:22469
IUPAC Name 2-(4-methylcyclohex-3-en-1-yl)propan-2-ol
SMILES CC1=CCC(CC1)C(C)(C)O

Specifications

Color White-Yellow
Boiling Point 218°C
Quantity 500 mL
Formula Weight 154.25
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material alpha-Terpineol
EINECSNumber (3)-2323
RTECSNumber WZ6700000
TSCA Yes