missing translation for 'onlineSavingsMsg'
Learn More

alpha-Phellandrene, Spectrum™ Chemical

Catalog No. 18600445
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600445 500 g Amber Glass Bottle
18600444 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18600445 Supplier Spectrum Chemical Mfg Cor Supplier No. P2590500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

P2590, 99-83-2, C10H16

Spectrum™ Chemical alpha-Phellandrene

Chemical Identifiers

CAS 99-83-2
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
InChI Key OGLDWXZKYODSOB-SNVBAGLBSA-N
IUPAC Name (5R)-2-methyl-5-(propan-2-yl)cyclohexa-1,3-diene
SMILES CC(C)[C@H]1CC=C(C)C=C1

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 500 g
UN Number UN2319
Grade Reagent