missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-naphtholphtaléine 98,0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 240°C |
| Quantity | 1 g |
| Formula Weight | 418.45 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Poudre cristalline |
| Chemical Name or Material | alpha-Naphtholphthalein |
Sécurité et manipulation
| EINECSNumber | (5)-3350 |
| TSCA | No |