missing translation for 'onlineSavingsMsg'
Learn More

alpha-Naphthoflavone, 97%

Catalog No. AC128160050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC128160050 5 g Glass bottle
AC128160010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC128160050 Supplier Thermo Scientific Chemicals Supplier No. 128160050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 604-59-1 | C19H12O2 | 272.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 604-59-1
Molecular Formula C19H12O2
Molecular Weight (g/mol) 272.29
MDL Number MFCD00004985
InChI Key VFMMPHCGEFXGIP-UHFFFAOYSA-N
Synonym alpha-naphthoflavone, 7,8-benzoflavone, 2-phenyl-4h-benzo h chromen-4-one, alpha-naphthylflavone, 2-phenylbenzo h chromen-4-one, benzo h flavone, 4h-naphtho 1,2-b pyran-4-one, 2-phenyl, .alpha.-naphthoflavone, 2-phenyl-benzo h chromen-4-one, 2-phenyl-4h-naphtho 1,2-b pyran-4-one
PubChem CID 11790
ChEBI CHEBI:76995
IUPAC Name 2-phenylbenzo[h]chromen-4-one
SMILES C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C4=CC=CC=C4C=C3

Specifications

CAS Min % 99.5
CAS Max % 100.0
Melting Point 155°C to 158°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 17, IV, 5551
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in ethanol,chloroform,soluble in sulfuric acid
Formula Weight 272.29
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material α-Naphthoflavone, 99.5+%
Show More Show Less
EINECSNumber 210-071-1
RTECSNumber QL6250000
TSCA TSCA

RUO – Research Use Only