missing translation for 'onlineSavingsMsg'
Learn More

alpha-Methylstyrene (stabilized with TBC) 99.0+%, TCI America™

Catalog No. M042925ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M042925ML 25 mL
M0429500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M042925ML Supplier TCI America Supplier No. M042925ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 98-83-9
Molecular Formula C9H10
Molecular Weight (g/mol) 118.18
MDL Number MFCD00008859
InChI Key XYLMUPLGERFSHI-UHFFFAOYSA-N
Synonym alpha-methylstyrene, 2-phenylpropene, 2-phenyl-1-propene, isopropenylbenzene, benzene, 1-methylethenyl, 2-phenylpropylene, 1-methyl-1-phenylethylene, alpha-methylstyrol, as-methylphenylethylene, 1-phenyl-1-methylethylene
PubChem CID 7407
ChEBI CHEBI:35060
IUPAC Name (prop-1-en-2-yl)benzene
SMILES CC(=C)C1=CC=CC=C1

Specifications

Melting Point -23°C
Color Colorless
Boiling Point 162°C
Quantity 25 mL
UN Number 2303
Formula Weight 118.18
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material alpha-Methylstyrene (stabilized with TBC)
EINECSNumber (3)-0005&(3)-0008
RTECSNumber WL5075300
TSCA Yes
Recommended Storage Refrigerator