missing translation for 'onlineSavingsMsg'
Learn More

alpha-Methylcinnamic acid, 99%

Catalog No. AAL0080622
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0080622 100 g
AAL0080606 5 g
AAL0080614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0080622 Supplier Thermo Scientific Chemicals Supplier No. L0080622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1199-77-5 | C10H10O2 | 162.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1199-77-5
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00002652
InChI Key XNCRUNXWPDJHGV-BQYQJAHWSA-N
Synonym alpha-methylcinnamic acid, 2-methyl-3-phenylacrylic acid, 2e-2-methyl-3-phenylprop-2-enoic acid, a-methylcinnamic acid, 2e-2-methyl-3-phenylacrylic acid, .alpha.-methylcinnamic acid, e-2-methyl-3-phenylacrylic acid, 2-propenoic acid, 2-methyl-3-phenyl, alpha-methyl cinnamic acid, e-2-methyl-3-phenylprop-2-enoic acid
PubChem CID 637817
IUPAC Name (E)-2-methyl-3-phenylprop-2-enoic acid
SMILES C\C(=C/C1=CC=CC=C1)C(O)=O

Specifications

Melting Point 78°C to 82°C
Boiling Point 288°C
Quantity 100 g
Beilstein 2042544
Formula Weight 162.19
Percent Purity 99%
Chemical Name or Material alpha-Methylcinnamic acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-847-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only